Tisk…

Anti-HNRNPM Mouse Monoclonal Antibody [clone: 2A6]

Dodavatel: OriGene
ORIGTA301557EA 16040 CZK
ORIGTA301557
Anti-HNRNPM Mouse Monoclonal Antibody [clone: 2A6]
Protilátky
  • Recommended Dilutions: Western Blot: 1: 1000-1: 10000

Type: Primary
Antigen: HNRNPM (heterogeneous nuclear ribonucleoprotein M)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR