Tisk…

Anti-ROBO4 Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA301644EA 16040 CZK
ORIGTA301644
Anti-ROBO4 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: ROBO4 (roundabout, axon guidance receptor, homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR