Tisk…
Anti-DMC1 Mouse Monoclonal Antibody [clone: 1D12/4]
Dodavatel: OriGene
ORIGTA301671EA 17590 CZK
ORIGTA301671
Anti-DMC1 Mouse Monoclonal Antibody [clone: 1D12/4]
Protilátky
Recommended Dilutions: Western Blot: 1: 1000
Type: Primary
Antigen: DMC1 (DNA meiotic recombinase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse