Tisk…

Anti-PARP4 Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA313436EA 15900 CZK
ORIGTA313436
Anti-PARP4 Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Immunochemistry: 1:50-1:100; ELISA: 1:10000

Type: Primary
Antigen: PARP4 (poly (ADP-ribose) polymerase family, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR