Tisk…

Anti-ALDOA Rabbit Polyclonal Antibody

Dodavatel: OriGene
ORIGTA313445EA 15900 CZK
ORIGTA313445
Anti-ALDOA Rabbit Polyclonal Antibody
Protilátky
Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000

Type: Primary
Antigen: ALDOA (aldolase A, fructose-bisphosphate)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR