Tisk…
Anti-FGF2 Mouse Monoclonal Antibody [clone: OTI2H11]
Dodavatel: OriGene
ORIGTA500013EA 17370 CZK
ORIGTA500013
Anti-FGF2 Mouse Monoclonal Antibody [clone: OTI2H11]
Protilátky
Recommended Dilutions: Western Blot: 1:5000-10000
Type: Primary
Antigen: FGF2 (fibroblast growth factor 2 (basic))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human