Anti-DGCR6L Rabbit polyclonal antibody
: Biorbyt
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)
Type: Primary
Antigen: DGCR6L (DiGeorge syndrome critical region gene 6-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- BIRBORB2966-200
- BIRBORB2966-100
- Antigen symbol:
- DGCR6L
- DGCR6L
- Antigen name:
- DiGeorge syndrome critical region gene 6-like
- DiGeorge syndrome critical region gene 6-like
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide
- Synthetic peptide
- Form:
- liquid
- liquid
- Purification:
- purified by Protein A chromatography
- purified by Protein A chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
- : ORB2966-200: BIRBORB2966-200
Specifications
Antigen symbolDGCR6LAntigen nameDiGeorge syndrome critical region gene 6-likeEnvironmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesImmunogenSynthetic peptideFormliquidPurificationpurified by Protein A chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIce - : ORB2966-100: BIRBORB2966-100
Specifications
Antigen symbolDGCR6LAntigen nameDiGeorge syndrome critical region gene 6-likeEnvironmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesImmunogenSynthetic peptideFormliquidPurificationpurified by Protein A chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIce