Tisk…

Anti-FAM102B Rabbit Polyclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB4352-100EA 10860 CZK
BIRBORB4352-100
Anti-FAM102B Rabbit Polyclonal Antibody (FITC (Fluorescein))
Protilátky
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: FAM102B (family with sequence similarity 102, member B)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR