Tisk…

Anti-ADAM8 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB4376-200EA 14320 CZK
BIRBORB4376-200 BIRBORB4376-20 BIRBORB4376-5 BIRBORB4376-100
Anti-ADAM8 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: ADAM8 (ADAM metallopeptidase domain 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR