Tisk…

Anti-CASC3 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB4409-100EA 8800 CZK
BIRBORB4409-100 BIRBORB4409-200
Anti-CASC3 Rabbit polyclonal antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CASC3 (cancer susceptibility candidate 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR