Tisk…

Anti-GATA3 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB10699-200EA 0 CZK
BIRBORB10699-200
Anti-GATA3 Rabbit polyclonal antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: GATA3 (GATA binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR