Tisk…
Anti-SLC2A2 Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB10726-100EA 0 CZK
BIRBORB10726-100 BIRBORB10726-200
Anti-SLC2A2 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: SLC2A2 (solute carrier family 2 (facilitated glucose transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat