Tisk…

Anti-CSF2 Rabbit polyclonal antibody

Dodavatel: Biorbyt
BIRBORB10731-100EA 0 CZK
BIRBORB10731-100 BIRBORB10731-200
Anti-CSF2 Rabbit polyclonal antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CSF2 (colony stimulating factor 2 (granulocyte-macrophage))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR