Tisk…
Anti-GSK3B Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB10754-200EA 0 CZK
BIRBORB10754-200 BIRBORB10754-100
Anti-GSK3B Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: GSK3B (glycogen synthase kinase 3 beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat