Tisk…

Anti-ACAP1 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB18800-100EA 0 CZK
BIRBORB18800-100
Anti-ACAP1 Goat Polyclonal Antibody
Protilátky
ELISA: 1:32000; Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ACAP1 (ArfGAP with coiled-coil, ankyrin repeat and PH domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR