Tisk…

Anti-IgG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Biorbyt
BIRBORB22249-0.2EA 0 CZK
BIRBORB22249-0.2
Anti-IgG Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR