Tisk…

Anti-IgM Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22282-0.2EA 0 CZK
BIRBORB22282-0.2
Anti-IgM Mouse Monoclonal Antibody
Protilátky
Type: Secondary
Antigen: IgM
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR