Tisk…

Anti-IgA/IgG1/IgG2/IgM Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22320-1EA 0 CZK
BIRBORB22320-1
Anti-IgA/IgG1/IgG2/IgM Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgA/IgG1/IgG2/IgM (Goat) (goat IgA/IgG1/IgG2/IgM)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR