Tisk…

Anti-IgG Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22327-1EA 0 CZK
BIRBORB22327-1
Anti-IgG Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG (Guinea pig) (guinea pig IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR