Tisk…

Anti-Hamster Serum Proteins Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22333-1EA 0 CZK
BIRBORB22333-1
Anti-Hamster Serum Proteins Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: Hamster Serum Proteins
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR