Tisk…
Anti-Horse Serum Proteins Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB22339-1EA 0 CZK
BIRBORB22339-1
Anti-Horse Serum Proteins Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: Horse Serum Proteins
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat