Tisk…

Anti-IgG lambda Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22353-1EA 0 CZK
BIRBORB22353-1
Anti-IgG lambda Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: lambda light chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR