Tisk…

Anti-IgE Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22356-1EA 0 CZK
BIRBORB22356-1
Anti-IgE Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR