Tisk…
Anti-J Chain Of Dimeric IgA Rabbit Polyclonal Antibody
Dodavatel: Biorbyt
BIRBORB22363-0.5EA 0 CZK
BIRBORB22363-0.5
Anti-J Chain Of Dimeric IgA Rabbit Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: J Chain Of Dimeric IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Objednat