Tisk…

Anti-IgA/IgG1/IgG2/IgM Sheep Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22431-1EA 0 CZK
BIRBORB22431-1
Anti-IgA/IgG1/IgG2/IgM Sheep Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: Guinea Pig IgG1 IgG2 IgA IgM
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR