Tisk…

Anti-IgD Sheep Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22437-1EA 0 CZK
BIRBORB22437-1
Anti-IgD Sheep Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR