Tisk…

Anti-IgG4 Sheep Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22440-1EA 0 CZK
BIRBORB22440-1
Anti-IgG4 Sheep Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: IgG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR