Tisk…

Anti-Rat Serum Proteins Sheep Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22451-1EA 0 CZK
BIRBORB22451-1
Anti-Rat Serum Proteins Sheep Polyclonal Antibody
Protilátky
Type: Secondary
Antigen: Rat Serum Proteins
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR