Tisk…

Anti-RTCB Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22473-100EA 0 CZK
BIRBORB22473-100
Anti-RTCB Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: RTCB (RNA 2',3'-cyclic phosphate and 5'-OH ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR