Tisk…

Anti-NCAM1 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22474-100EA 0 CZK
BIRBORB22474-100
Anti-NCAM1 Goat Polyclonal Antibody
Protilátky
ELISA: 1:32000; Western Blot: 0.01-0.03 ?g/ml

Type: Primary
Antigen: NCAM1 (neural cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR