Tisk…

Anti-DNAJB9 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22490-100EA 0 CZK
BIRBORB22490-100
Anti-DNAJB9 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: DNAJB9 (DnaJ (Hsp40) homolog, subfamily B, member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR