Tisk…

Anti-IDH3A Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22494-100EA 0 CZK
BIRBORB22494-100
Anti-IDH3A Goat Polyclonal Antibody
Protilátky
ELISA: 1:32000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: IDH3A (isocitrate dehydrogenase 3 (NAD+) alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR