Tisk…

Anti-CAPE Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22507-100EA 0 CZK
BIRBORB22507-100
Anti-CAPE Goat Polyclonal Antibody
Protilátky
ELISA: 1:32000; Western Blot: 0.2-0.6 ?g/ml

Type: Primary
Antigen: CAPE
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR