Tisk…

Anti-COPS3 Goat Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22517-100EA 0 CZK
BIRBORB22517-100
Anti-COPS3 Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: COPS3 (COP9 signalosome subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR