Tisk…

Anti-ANGPTL3 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22572-20EA 0 CZK
BIRBORB22572-20 BIRBORB22572-100 BIRBORB22572-5
Anti-ANGPTL3 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ANGPTL3 (angiopoietin-like 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR