Tisk…
Anti-BMP7 Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB22576-20EA 0 CZK
BIRBORB22576-20 BIRBORB22576-100 BIRBORB22576-5
Anti-BMP7 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: BMP7 (bone morphogenetic protein 7)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human