Tisk…

Anti-PTPRC Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22669-20EA 0 CZK
BIRBORB22669-20 BIRBORB22669-100 BIRBORB22669-5
Anti-PTPRC Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR