Tisk…

Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein))

Dodavatel: Biorbyt
BIRBORB22675-500EA 0 CZK
BIRBORB22675-500 BIRBORB22675-1
Anti-CD58 Mouse Monoclonal Antibody (FITC (Fluorescein))
Protilátky
Type: Primary
Antigen: CD58 (CD58 molecule)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR