Tisk…

Anti-CD8A Rat Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB22693-500EA 0 CZK
BIRBORB22693-500 BIRBORB22693-1
Anti-CD8A Rat Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CD8A (CD8a molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgM (Rat)
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR