Tisk…

Anti-CD28 Hamster Monoclonal Antibody (Biotin)

Dodavatel: Biorbyt
BIRBORB22696-1EA 0 CZK
BIRBORB22696-1 BIRBORB22696-500
Anti-CD28 Hamster Monoclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Hamster
Isotype: IgG (Hamster)
Reactivity: Mouse
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR