Tisk…

Anti-Influenza Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22723-10EA 0 CZK
BIRBORB22723-10 BIRBORB22723-50
Anti-Influenza Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Influenza
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Virus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR