Tisk…

Anti-Influenza H1N1 Rabbit Polyclonal Antibody

Dodavatel: Biorbyt
BIRBORB22735-100EA 0 CZK
BIRBORB22735-100 BIRBORB22735-15 BIRBORB22735-5
Anti-Influenza H1N1 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Influenza H1N1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Virus
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR