Tisk…

Anti-CRYAB Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB22741-5EA 0 CZK
BIRBORB22741-5 BIRBORB22741-100 BIRBORB22741-20
Anti-CRYAB Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR