Tisk…
Anti-ST13 Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB22750-20EA 0 CZK
BIRBORB22750-20 BIRBORB22750-100 BIRBORB22750-5
Anti-ST13 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: ST13 (suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein))
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human