Tisk…
Anti-AGXT Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB22752-5EA 0 CZK
BIRBORB22752-5 BIRBORB22752-100 BIRBORB22752-20
Anti-AGXT Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: AGXT (alanine-glyoxylate aminotransferase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human