Tisk…
Anti-CTSL Mouse Monoclonal Antibody
Dodavatel: Biorbyt
BIRBORB22765-5EA 0 CZK
BIRBORB22765-5 BIRBORB22765-100 BIRBORB22765-20
Anti-CTSL Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: CTSL (cathepsin L)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human