Tisk…

Anti-Cyclophilin Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22766-5EA 0 CZK
BIRBORB22766-5 BIRBORB22766-100 BIRBORB22766-20
Anti-Cyclophilin Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: Cyclophilin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR