Tisk…

Anti-FKBP4 Mouse monoclonal antibody

Dodavatel: Biorbyt
BIRBORB22768-100EA 0 CZK
BIRBORB22768-100 BIRBORB22768-20 BIRBORB22768-5
Anti-FKBP4 Mouse monoclonal antibody
Protilátky
Type: Primary
Antigen: FKBP4 (FK506 binding protein 4, 59kDa)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR