Tisk…

Anti-HTRA2 Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22774-5EA 0 CZK
BIRBORB22774-5 BIRBORB22774-20 BIRBORB22774-100
Anti-HTRA2 Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: HTRA2 (HtrA serine peptidase 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR