Tisk…

Anti-PPM1A Mouse Monoclonal Antibody

Dodavatel: Biorbyt
BIRBORB22782-5EA 0 CZK
BIRBORB22782-5 BIRBORB22782-100 BIRBORB22782-20
Anti-PPM1A Mouse Monoclonal Antibody
Protilátky
Type: Primary
Antigen: PPM1A (protein phosphatase, Mg2+/Mn2+ dependent, 1A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b (Mouse)
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR